siRNA Details
| Gene Symbol: OR56B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005180.2 | NP_001005180.1 | 1 | 981 | ||
| GenBank mRNA | AK122668.1 | - | 1 | 981 | ||
| BC137184.1 | - | 1 | 978 | 9021181 | ||
| BC137185.1 | - | 1 | 978 | 9021182 | ||

OR56B1
Genomic Map