siRNA Details
| Gene Symbol: LOC101929584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120476.2 | 3 | 378 | |||
| NR_120477.1 | 3 | 378 | ||||
| GenBank mRNA | BC031884.1 | - | 3 | 389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929584| Gene Symbol: LOC101929584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120476.2 | 3 | 378 | |||
| NR_120477.1 | 3 | 378 | ||||
| GenBank mRNA | BC031884.1 | - | 3 | 389 | ||