siRNA Details
Gene Symbol: LOC101929584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120476.2 | 3 | 363 | |||
NR_120477.1 | 3 | 363 | ||||
GenBank mRNA | BC031884.1 | - | 3 | 374 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120476.2 | 3 | 363 | |||
NR_120477.1 | 3 | 363 | ||||
GenBank mRNA | BC031884.1 | - | 3 | 374 |