siRNA Details
Gene Symbol: SERP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010897.1 | NP_001010897.1 | 3 | 718 | ||
GenBank mRNA | AK123212.1 | - | 1 | 1518 | ||
BC029067.1 | - | 3 | 718 | 5197847 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SERP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010897.1 | NP_001010897.1 | 3 | 718 | ||
GenBank mRNA | AK123212.1 | - | 1 | 1518 | ||
BC029067.1 | - | 3 | 718 | 5197847 |