siRNA Details
| Gene Symbol: LOC107984562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020896.1 | XP_016876385.1 | 2, 3 | 782 | ||
| XM_017020897.1 | XP_016876386.1 | 3, 4 | 367 | |||
| XM_017020898.1 | XP_016876387.1 | 3, 4 | 370 | |||

LOC107984562
Genomic Map