siRNA Details
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001444.2 | NP_001435.1 | 4 | 697 | ||
GenBank mRNA | AK226036.1 | - | 4 | 646 | ||
BC019385.2 | - | 4 | 630 | 3862100 | ||
BC070303.1 | - | 1 | 644 | 6496304 | ||
M94856.1 | - | 4 | 632 |
