siRNA Details
| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001444.2 | NP_001435.1 | 4 | 700 | ||
| GenBank mRNA | AK226036.1 | - | 4 | 649 | ||
| BC019385.2 | - | 4 | 633 | 3862100 | ||
| BC070303.1 | - | 1 | 647 | 6496304 | ||
| M94856.1 | - | 4 | 635 | |||

FABP5
Genomic Map