siRNA Details
| Gene Symbol: FABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001444.2 | NP_001435.1 | 2 | 207 | ||
| GenBank mRNA | AK226036.1 | - | 2 | 156 | ||
| AK311856.1 | - | 2 | 154 | |||
| BC019385.2 | - | 2 | 140 | 3862100 | ||
| BC070303.1 | - | 1 | 154 | 6496304 | ||
| BT007449.1 | - | 1 | 94 | |||
| CN278427.1 | - | 2 | 327 | |||
| M94856.1 | - | 2 | 142 | |||

FABP5
Genomic Map