siRNA Details
| Gene Symbol: LINC01551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026732.1 | 3 | 293 | |||
| GenBank mRNA | BM839073.1 | - | 3 | 212 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01551| Gene Symbol: LINC01551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026732.1 | 3 | 293 | |||
| GenBank mRNA | BM839073.1 | - | 3 | 212 | ||