siRNA Details
| Gene Symbol: TOMM20L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207377.2 | NP_997260.1 | 1 | 117 | ||
| XM_011536742.2 | XP_011535044.1 | 1 | 136 | |||
| XM_011536743.2 | XP_011535045.1 | 1 | 139 | |||
| XM_011536744.2 | XP_011535046.1 | 1 | 137 | |||
| GenBank mRNA | AY358266.1 | - | 1 | 129 | ||
| BC137557.1 | - | 1 | 115 | 9021557 | ||
| BC137563.1 | - | 1 | 115 | 9021563 | ||
| BC144709.1 | - | 1 | 115 | |||

TOMM20L
Genomic Map