siRNA Details
| Gene Symbol: C15orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039614.2 | NP_001034703.1 | 2 | 660 | ||
| NM_001303254.1 | NP_001290183.1 | 3 | 543 | |||
| XM_005254369.4 | XP_005254426.1 | 3 | 1943 | |||
| GenBank mRNA | BC111368.1 | - | 2 | 660 | 7504115 | |
| DR003635.1 | - | 3 | 550 | |||
| DY655797.1 | - | 2 | 370 | |||

C15orf59
Genomic Map