siRNA Details
| Gene Symbol: UBE2Q2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243531.1 | NP_001230460.1 | 3 | 455 | ||
| GenBank mRNA | CB050071.1 | - | 3 | 461 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2Q2L| Gene Symbol: UBE2Q2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243531.1 | NP_001230460.1 | 3 | 455 | ||
| GenBank mRNA | CB050071.1 | - | 3 | 461 | ||