siRNA Details
| Gene Symbol: C16orf87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001436.2 | NP_001001436.1 | 2 | 97 | ||
| GenBank mRNA | BC056676.1 | - | 2 | 101 | 6711353 | |
| BX647945.1 | - | 2 | 97 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf87| Gene Symbol: C16orf87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001436.2 | NP_001001436.1 | 2 | 97 | ||
| GenBank mRNA | BC056676.1 | - | 2 | 101 | 6711353 | |
| BX647945.1 | - | 2 | 97 | |||