siRNA Details
| Gene Symbol: C16orf86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012984.2 | NP_001013002.2 | 3 | 691 | ||
| XM_005255952.4 | XP_005256009.1 | 3 | 699 | |||
| GenBank mRNA | AK123796.1 | - | 2 | 1041 | ||
| BC032064.1 | - | 3 | 668 | |||
| BC064498.1 | - | 3 | 737 | |||
| BC082249.1 | - | 3 | 737 | |||
| BC132878.1 | - | 1 | 606 | |||
| BC132880.1 | - | 1 | 606 | |||

C16orf86
Genomic Map