siRNA Details
Gene Symbol: C17orf98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080465.2 | NP_001073934.1 | 3 | 371 | ||
GenBank mRNA | DY654789.1 | - | 3 | 322 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C17orf98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080465.2 | NP_001073934.1 | 3 | 371 | ||
GenBank mRNA | DY654789.1 | - | 3 | 322 |