siRNA Details
| Gene Symbol: RPRML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203400.4 | NP_981945.1 | 1 | 545 | ||
| GenBank mRNA | BC033942.1 | - | 1 | 550 | 5274621 | |
| BC108690.1 | - | 1 | 520 | 6140781 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPRML| Gene Symbol: RPRML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203400.4 | NP_981945.1 | 1 | 545 | ||
| GenBank mRNA | BC033942.1 | - | 1 | 550 | 5274621 | |
| BC108690.1 | - | 1 | 520 | 6140781 | ||