siRNA Details
| Gene Symbol: BTBD17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080466.1 | NP_001073935.1 | 3 | 1363 | ||
| XM_011524791.2 | XP_011523093.1 | 5 | 2269 | |||
| XM_017024622.1 | XP_016880111.1 | 5 | 2205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BTBD17| Gene Symbol: BTBD17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080466.1 | NP_001073935.1 | 3 | 1363 | ||
| XM_011524791.2 | XP_011523093.1 | 5 | 2269 | |||
| XM_017024622.1 | XP_016880111.1 | 5 | 2205 | |||