siRNA Details
| Gene Symbol: CLEC17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204118.1 | NP_001191047.1 | 9 | 651 | ||
| NM_207390.3 | NP_997273.3 | 9 | 651 | |||
| NR_109784.1 | 9 | 651 | ||||
| NR_109785.1 | 9 | 651 | ||||
| XM_017026785.1 | XP_016882274.1 | 10 | 1195 | |||
| XM_017026786.1 | XP_016882275.1 | 10 | 1059 | |||
| XM_017026787.1 | XP_016882276.1 | 11 | 1414 | |||
| XM_017026788.1 | XP_016882277.1 | 10 | 1171 | |||
| XM_017026789.1 | XP_016882278.1 | 6 | 873 | |||
| XM_017026790.1 | XP_016882279.1 | 9 | 1147 | |||
| XM_017026792.1 | XP_016882281.1 | 10 | 1199 | |||
| XM_017026793.1 | XP_016882282.1 | 10 | 1202 | |||
| XM_017026794.1 | XP_016882283.1 | 10 | 1204 | |||
| GenBank mRNA | BC140848.1 | - | 9 | 612 | 9021716 | |
| BC144664.1 | - | 9 | 612 | |||
| BC144665.1 | - | 9 | 612 | 9053196 | ||

CLEC17A
Genomic Map