siRNA Details
| Gene Symbol: KRTDAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244847.1 | NP_001231776.1 | 4, 5 | 304 | ||
| NM_207392.2 | NP_997275.1 | 5, 6 | 346 | |||
| GenBank mRNA | AY359007.1 | - | 5, 6 | 269 | ||
| BC130501.1 | - | 5, 6 | 346 | 40146532 | ||
| W95920.1 | - | 4, 5 | 213 | |||

KRTDAP
Genomic Map