siRNA Details
| Gene Symbol: ZNF790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242800.1 | NP_001229729.1 | 5 | 882 | ||
| NM_001242801.1 | NP_001229730.1 | 5 | 1060 | |||
| NM_001242802.1 | NP_001229731.1 | 5 | 1150 | |||
| NM_206894.3 | NP_996777.2 | 5 | 842 | |||
| XM_005258903.4 | XP_005258960.1 | 5 | 1209 | |||
| XM_011526950.2 | XP_011525252.1 | 5 | 1290 | |||
| GenBank mRNA | AK297474.1 | - | 5 | 851 | ||
| BC057245.1 | - | 5 | 844 | 6712908 | ||
| BM473790.1 | - | 3 | 682 | |||

ZNF790
Genomic Map