siRNA Details
| Gene Symbol: ZNF761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008401.3 | NP_001008401.3 | 6 | 1174 | ||
| NM_001289951.1 | NP_001276880.1 | 5 | 955 | |||
| NM_001289952.1 | NP_001276881.1 | 5 | 1023 | |||
| GenBank mRNA | AB107355.2 | - | 5 | 1024 | ||
| AK131281.1 | - | 2 | 644 | |||

ZNF761
Genomic Map