siRNA Details
| Gene Symbol: ZNF749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001023561.3 | NP_001018855.2 | 3 | 1359 | ||
| NM_001321952.1 | NP_001308881.1 | 3 | 1433 | |||
| NM_001321953.1 | NP_001308882.1 | 2 | 1306 | |||
| NM_001321954.1 | NP_001308883.1 | 2 | 1232 | |||
| XM_011526956.2 | XP_011525258.1 | 3 | 1108 | |||
| XM_017026802.1 | XP_016882291.1 | 4 | 2359 | |||
| GenBank mRNA | AK122740.1 | - | 1 | 101 | ||
| AK122794.1 | - | 3 | 1355 | |||

ZNF749
Genomic Map