siRNA Details
Gene Symbol: ZNF749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023561.3 | NP_001018855.2 | 3 | 1486 | ||
NM_001321952.1 | NP_001308881.1 | 3 | 1560 | |||
NM_001321953.1 | NP_001308882.1 | 2 | 1433 | |||
NM_001321954.1 | NP_001308883.1 | 2 | 1359 | |||
XM_011526956.2 | XP_011525258.1 | 3 | 1235 | |||
XM_017026802.1 | XP_016882291.1 | 4 | 2486 | |||
GenBank mRNA | AK122740.1 | - | 1 | 228 | ||
AK122794.1 | - | 3 | 1482 |
