siRNA Details
| Gene Symbol: ZNF324B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207395.2 | NP_997278.2 | 4 | 2057 | ||
| XM_005258915.4 | XP_005258972.1 | 5 | 2130 | |||
| XM_005258918.4 | XP_005258975.1 | 4 | 2081 | |||
| GenBank mRNA | AK127750.1 | - | 4 | 2057 | ||
| AK131452.1 | - | 3 | 2852 | |||
| BC140881.1 | - | 4 | 2050 | 9021749 | ||
| BC140882.1 | - | 5 | 2130 | 9021750 | ||
| BX648711.1 | - | 4 | 2030 | |||

ZNF324B
Genomic Map