siRNA Details
| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207396.2 | NP_997279.2 | 15 | 1670 | ||
| XM_011541439.2 | XP_011539741.1 | 14 | 2191 | |||
| XM_017001259.1 | XP_016856748.1 | 15 | 1690 | |||
| GenBank mRNA | AK056658.1 | - | 8 | 881 | ||
| AK127700.1 | - | 3 | 325 | |||
| AK294223.1 | - | 10 | 985 | |||
| BC128237.1 | - | 9 | 879 | |||

RNF207
Genomic Map