siRNA Details
| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207396.2 | NP_997279.2 | 11 | 1174 | ||
| XM_011541439.2 | XP_011539741.1 | 10 | 1695 | |||
| XM_017001259.1 | XP_016856748.1 | 11 | 1128 | |||
| GenBank mRNA | AK294223.1 | - | 6 | 489 | ||
| BC128237.1 | - | 5 | 383 | |||
| DA190055.1 | - | 5 | 337 | |||

RNF207
Genomic Map