siRNA Details
Gene Symbol: TRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013642.2 | NP_001013664.1 | 1 | 206 | ||
XM_005245867.3 | XP_005245924.1 | 1 | 247 | |||
GenBank mRNA | BC069216.1 | - | 1 | 203 | 6376829 | |
CK904126.1 | - | 1 | 206 |
