siRNA Details
| Gene Symbol: CD164L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207397.3 | NP_997280.2 | 5 | 1132 | ||
| GenBank mRNA | AY358761.1 | - | 5 | 1102 | ||
| BC137466.1 | - | 5 | 1091 | 9021466 | ||
| BM145906.1 | - | 1 | 43 | |||
| BM145910.1 | - | 1 | 43 | |||

CD164L2
Genomic Map