siRNA Details
Gene Symbol: SLC6A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010898.2 | NP_001010898.1 | 7 | 1512 | ||
XM_006710643.2 | XP_006710706.1 | 7 | 1220 | |||
GenBank mRNA | AK289982.1 | - | 7 | 1508 | ||
BC140908.1 | - | 6 | 1097 | 9056720 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
