siRNA Details
| Gene Symbol: C1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010979.2 | NP_001010979.1 | 4 | 316 | ||
| GenBank mRNA | BC127710.1 | - | 4 | 315 | 40127224 | |
| BC130509.1 | - | 4 | 374 | 40146540 | ||
| HY025946.1 | - | 4 | 316 | |||

C1orf189
Genomic Map