siRNA Details
| Gene Symbol: CAPN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143962.1 | NP_001137434.1 | 9 | 1130 | ||
| XM_017001265.1 | XP_016856754.1 | 6 | 571 | |||
| XM_017001266.1 | XP_016856755.1 | 4 | 488 | |||
| XM_017001267.1 | XP_016856756.1 | 9 | 1130 | |||
| XM_017001268.1 | XP_016856757.1 | 9 | 1130 | |||
| GenBank mRNA | BC157893.1 | - | 9 | 1067 | 9057098 | |

CAPN8
Genomic Map