siRNA Details
| Gene Symbol: COA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012985.2 | NP_001013003.1 | 3 | 348 | ||
| NM_001206641.2 | NP_001193570.2 | 3 | 432 | |||
| NM_001301733.1 | NP_001288662.1 | 2 | 719 | |||
| GenBank mRNA | BC025793.1 | - | 3 | 255 | ||
| BC035925.1 | - | 3 | 345 | |||
| BC116455.1 | - | 3 | 348 | 4540930 | ||
| BG204581.1 | - | 2 | 191 | |||
| CB136291.1 | - | 3 | 401 | |||
| HY169996.1 | - | 3 | 435 | |||

COA6
Genomic Map