siRNA Details
| Gene Symbol: C1orf229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207401.2 | NP_997284.1 | 1 | 1478 | ||
| GenBank mRNA | AK127619.1 | - | 1 | 1478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf229| Gene Symbol: C1orf229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207401.2 | NP_997284.1 | 1 | 1478 | ||
| GenBank mRNA | AK127619.1 | - | 1 | 1478 | ||