siRNA Details
| Gene Symbol: LINC00493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF147354.1 | - | 1, 2 | 145 | ||
| BC039422.1 | - | 1, 2 | 146 | |||
| BC092494.1 | - | 1, 2 | 168 | |||
| BC127753.1 | - | 1, 2 | 146 | |||
| BC127754.1 | - | 1, 2 | 146 | |||
| BC141963.1 | - | 1, 2 | 146 | |||
| BC141964.1 | - | 1, 2 | 146 | |||

LINC00493
Genomic Map