siRNA Details
| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 3, 4 | 266 | |||
| NR_028411.1 | 2, 3 | 199 | ||||
| GenBank mRNA | BC144508.1 | - | 3, 4 | 254 | ||
| BU585030.1 | - | 2, 3 | 218 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00207| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 3, 4 | 266 | |||
| NR_028411.1 | 2, 3 | 199 | ||||
| GenBank mRNA | BC144508.1 | - | 3, 4 | 254 | ||
| BU585030.1 | - | 2, 3 | 218 | |||