siRNA Details
| Gene Symbol: TMEM247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145051.2 | NP_001138523.1 | 3 | 583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM247| Gene Symbol: TMEM247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145051.2 | NP_001138523.1 | 3 | 583 | ||