siRNA Details
| Gene Symbol: C2orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080474.1 | NP_001073943.1 | 2 | 426 | ||
| XM_005264302.3 | XP_005264359.1 | 2 | 371 | |||
| GenBank mRNA | AK125975.1 | - | 2 | 426 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf78| Gene Symbol: C2orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080474.1 | NP_001073943.1 | 2 | 426 | ||
| XM_005264302.3 | XP_005264359.1 | 2 | 371 | |||
| GenBank mRNA | AK125975.1 | - | 2 | 426 | ||