siRNA Details
| Gene Symbol: SP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003845.2 | NP_001003845.1 | 2 | 1289 | ||
| XM_005246542.4 | XP_005246599.1 | 2 | 2416 | |||
| XM_011511159.2 | XP_011509461.1 | 2 | 2452 | |||
| GenBank mRNA | AB096175.1 | - | 2 | 1285 | ||
| BQ064677.1 | - | 1 | 173 | |||

SP5
Genomic Map