siRNA Details
| Gene Symbol: PLEKHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080475.2 | NP_001073944.1 | 2 | 533 | ||
| XM_005246545.3 | XP_005246602.1 | 2 | 533 | |||
| XM_006712521.3 | XP_006712584.1 | 2 | 533 | |||
| XM_011511160.2 | XP_011509462.1 | 3 | 868 | |||
| XM_011511163.2 | XP_011509465.1 | 2 | 533 | |||
| XM_017004071.1 | XP_016859560.1 | 2 | 533 | |||
| XM_017004073.1 | XP_016859562.1 | 2 | 533 | |||
| GenBank mRNA | BC151137.1 | - | 1 | 172 | 9057037 | |
| BX648983.1 | - | 2 | 472 | |||

PLEKHM3
Genomic Map