siRNA Details
| Gene Symbol: FOXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005250.2 | NP_005241.1 | 1 | 615 | ||
| GenBank mRNA | BC100027.1 | - | 1 | 615 | 30920793 | |
| BC117226.1 | - | 1 | 443 | 40125777 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008024, BC137806, AB221616, BC137805.

FOXL1
Genomic Map