siRNA Details
Gene Symbol: ZNF662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134656.1 | NP_001128128.1 | 4 | 865 | ||
NM_207404.3 | NP_997287.2 | 5 | 953 | |||
GenBank mRNA | AK127779.1 | - | 5 | 953 | ||
AK128676.1 | - | 3 | 1504 | |||
AK131235.1 | - | 1 | 403 | |||
BC128085.1 | - | 3 | 578 | 40108370 |
