siRNA Details
| Gene Symbol: PRR23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013650.2 | NP_001013672.1 | 1 | 1818 | ||
| GenBank mRNA | AK127998.1 | - | 1 | 1819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR23B| Gene Symbol: PRR23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013650.2 | NP_001013672.1 | 1 | 1818 | ||
| GenBank mRNA | AK127998.1 | - | 1 | 1819 | ||