siRNA Details
| Gene Symbol: PLSCR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085420.1 | NP_001078889.1 | 4 | 1375 | ||
| NM_001321245.1 | NP_001308174.1 | 4 | 1339 | |||
| XM_017006373.1 | XP_016861862.1 | 4 | 586 | |||
| GenBank mRNA | AY436642.1 | - | 4 | 1375 | ||
| BC157886.1 | - | 4 | 362 | 8862707 | ||
| BC171917.1 | - | 4 | 362 | 9054571 | ||

PLSCR5
Genomic Map