siRNA Details
| Gene Symbol: TEX43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207408.1 | NP_997291.1 | 1 | 69 | ||
| GenBank mRNA | AK131015.1 | - | 1 | 69 | ||
| BC130495.1 | - | 1 | 77 | 40146526 | ||
| BC130497.1 | - | 1 | 77 | 40146528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX43| Gene Symbol: TEX43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207408.1 | NP_997291.1 | 1 | 69 | ||
| GenBank mRNA | AK131015.1 | - | 1 | 69 | ||
| BC130495.1 | - | 1 | 77 | 40146526 | ||
| BC130497.1 | - | 1 | 77 | 40146528 | ||