siRNA Details
| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286550.1 | NP_001273479.1 | 1 | 100 | ||
| NM_207409.3 | NP_997292.2 | 1 | 100 | |||
| GenBank mRNA | AY358823.1 | - | 1 | 79 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLPSL2| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286550.1 | NP_001273479.1 | 1 | 100 | ||
| NM_207409.3 | NP_997292.2 | 1 | 100 | |||
| GenBank mRNA | AY358823.1 | - | 1 | 79 | ||