siRNA Details
| Gene Symbol: SAMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030060.2 | NP_001025231.1 | 1 | 301 | ||
| XM_017010850.1 | XP_016866339.1 | 1 | 284 | |||
| GenBank mRNA | CB988162.1 | - | 1 | 330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAMD5| Gene Symbol: SAMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030060.2 | NP_001025231.1 | 1 | 301 | ||
| XM_017010850.1 | XP_016866339.1 | 1 | 284 | |||
| GenBank mRNA | CB988162.1 | - | 1 | 330 | ||