siRNA Details
| Gene Symbol: SAMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030060.2 | NP_001025231.1 | 1 | 649 | ||
| XM_017010850.1 | XP_016866339.1 | 1 | 632 | |||
| GenBank mRNA | BX642659.1 | - | 1 | 17 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAMD5| Gene Symbol: SAMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030060.2 | NP_001025231.1 | 1 | 649 | ||
| XM_017010850.1 | XP_016866339.1 | 1 | 632 | |||
| GenBank mRNA | BX642659.1 | - | 1 | 17 | ||