siRNA Details
Gene Symbol: LOC100287846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037168.1 | 1 | 199 | |||
GenBank mRNA | DB469678.1 | - | 1 | 200 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100287846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037168.1 | 1 | 199 | |||
GenBank mRNA | DB469678.1 | - | 1 | 200 |