siRNA Details
| Gene Symbol: LOC100287846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037168.1 | 1 | 142 | |||
| GenBank mRNA | DB469678.1 | - | 1 | 142 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100287846| Gene Symbol: LOC100287846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037168.1 | 1 | 142 | |||
| GenBank mRNA | DB469678.1 | - | 1 | 142 | ||