siRNA Details
| Gene Symbol: FAM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207413.3 | NP_997296.1 | 4 | 547 | ||
| GenBank mRNA | AY358213.1 | - | 4 | 550 | ||
| BC130641.1 | - | 4 | 494 | 40146672 | ||
| BC130643.1 | - | 4 | 494 | 40146674 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM150A| Gene Symbol: FAM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207413.3 | NP_997296.1 | 4 | 547 | ||
| GenBank mRNA | AY358213.1 | - | 4 | 550 | ||
| BC130641.1 | - | 4 | 494 | 40146672 | ||
| BC130643.1 | - | 4 | 494 | 40146674 | ||